Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf50 Monoclonal Antibody (OTI1F9)
GREENER_CHOICE

Catalog No. PIMA525542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525542 Supplier Invitrogen™ Supplier No. MA525542
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C1orf50 Monoclonal Antibody for Western Blot, ICC/IF, Flow

C1orf50 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf50
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1F9
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C1orf50
Gene Accession No. Q9BV19
Gene Alias C15H1orf50; C1orf50; chromosome 1 open reading frame 50; chromosome 15 C1orf50 homolog; uncharacterized protein C1orf50; uncharacterized protein C1orf50 homolog
Gene Symbols C1orf50
Host Species Mouse
Immunogen Full length human recombinant protein of C1orf50 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79078
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.