Learn More
C1orf50 Mouse anti-Human, Clone: OTI4D10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502938

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C1orf50 is a protein coding gene.Specifications
C1orf50 | |
Monoclonal | |
Unconjugated | |
C1orf50 | |
chromosome 1 open reading frame 50 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79078 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BV19 | |
C1orf50 | |
Full length human recombit protein of human C1orf50 produced in HEK293 cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.