Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111391 Supplier Invitrogen™ Supplier No. PA5111391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLGGGKHQLT KHFPSHHSDS AASSPASPME KMDQTQLGHL ALKPKQPWHL TQWPAMNLTW IHTTPICNPP LSSPGTISFS HGPLGTGTGI Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 72%.

Transcriptional repressor which forms a negative regulatory component of the circadian clock and acts independently of the circadian transcriptional repressors: CRY1, CRY2 and BHLHE41. In a histone deacetylase-dependent manner represses the transcriptional activator activity of the CLOCK-ARNTL/BMAL1 heterodimer. Abrogates the interaction of ARNTL/BMAL1 with the transcriptional coactivator CREBBP and can repress the histone acetyl-transferase activity of the CLOCK-ARNTL/BMAL1 heterodimer, reducing histone acetylation of its target genes. Rhythmically binds the E-box elements (5'-CACGTG-3') on circadian gene promoters and its occupancy shows circadian oscillation antiphasic to ARNTL/BMAL1. Interacts with the glucocorticoid receptor (NR3C1) and contributes to the repressive function in the glucocorticoid response. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf51
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ciart
Gene Accession No. Q8N365
Gene Alias C1orf51; C5H1orf51; ChIP-derived repressor of network oscillator; CHRONO; Ciart; circadian associated repressor of transcription; circadian-associated transcriptional repressor; Computationally highlighted repressor of the network oscillator; GM129; hypothetical gene supported by NM_017187; RGD1566380
Gene Symbols Ciart
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human C1orf51. Recombinant protein control fragment (Product #RP-106532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.