Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf57 Polyclonal Antibody
C1orf57 Polyclonal Antibody
Supplier: Thermo Scientific PA575971
Description
C1orf57 Polyclonal Antibody for Western Blot
Specifications
| C1orf57 | |
| Polyclonal | |
| Unconjugated | |
| Ntpcr | |
| 2310079N02Rik; AI449709; C1orf57; cancer-related nucleoside-triphosphatase; Cancer-related nucleoside-triphosphatase homolog; CH1073-266P11.1; HCR-NTPase; NTPase; Ntpcr; Nucleoside triphosphate phosphohydrolase; nucleoside-triphosphatase C1orf57 homolog; nucleoside-triphosphatase, cancer-related; RGD1306192; RP4-659I19.2; RP4-678E16.2; zgc:92420 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361443, 66566, 84284 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BSD7, Q9CQA9 | |
| Ntpcr | |
| Synthetic peptide corresponding to amino acids 145-189 of Human C1orf57. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction