Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf58 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598496
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598496 Supplier Invitrogen™ Supplier No. PA598496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf58 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

BROX (BRO1 Domain And CAAX Motif Containing) is a Protein Coding gene with functional area in the extracellular exosome but its exact function is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf58
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BROX
Gene Accession No. Q5VW32
Gene Alias BRO1 domain and CAAX motif containing; BRO1 domain- and CAAX motif-containing protein; BRO1 domain-containing protein BROX; BROFTI; BROX; C1orf58; RP11-452F19.1
Gene Symbols BROX
Host Species Rabbit
Immunogen Recombinant Human BRO1 domain-containing protein BROX protein (288-409AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148362
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.