Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf64 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3426 Supplier Bioss Supplier No. BS15073R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The estrogen receptor-related factor (ERRF) is a recently identified protein whose expression is upregulated in multiple cell lines and primary breast cancer tumors. Increased ERRF expression was significantly associated with ER or progesterone receptor (PR) positivity even in HER2-negative tumors. This higher ERRF expression correlated with better disease-free survival and overall survival. In two ER-positive breast cancer cell lines, knockdown of ERRF suppressed cell growth in vitro and tumorigenesis in xenograft models, suggesting that ERRF plays a role in estrogen-ER-mediated breast cancer cell growth and may thus be a potential therapeutic target.

Specifications

Antigen C1orf64
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SRARP
Gene Accession No. Q8NEQ6
Gene Alias C1orf64; chromosome 1 open reading frame 64; ER-related factor; ERRF; Gm694; predicted gene 694; RP11-5P18.4; Srarp; SSPR; Steroid receptor-associated and regulated protein; Steroid receptor-regulated protein; uncharacterized protein C1orf64; uncharacterized protein C1orf64 homolog
Gene Symbols SRARP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf64.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149563
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.