Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556498 Supplier Invitrogen™ Supplier No. PA556498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPVRFLDPLP TSGITPGATV LTASGQTVGK FRAGQGNVGL ALLWSEKIKG PLHIRASEGA QVALAASVPD WWPTVSK Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 65%.

Involved in the maturation of mitochondrial 4Fe-4S proteins functioning late in the iron-sulfur cluster assembly pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf69
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IBA57
Gene Accession No. Q5T440
Gene Alias 4930543L23Rik; A230051G13Rik; C1orf69; IBA57; IBA57 homolog, iron-sulfur cluster assembly; IBA57, iron-sulfur cluster assembly homolog; IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae); iron-sulfur cluster assembly factor for biotin synthase- and aconitase-like mitochondrial proteins, with a mass of 57kDa; iron-sulfur cluster assembly factor homolog; MMDS3; putative transferase C1orf69 homolog, mitochondrial; putative transferase C1orf69, mitochondrial; putative transferase CAF17 homolog, mitochondrial; Putative transferase CAF17, mitochondrial; RGD1306000; SPG74
Gene Symbols IBA57
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf69. Recombinant protein control fragment (Product #RP-104192).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.