Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598489
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598489 Supplier Invitrogen™ Supplier No. PA598489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf69 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Involved in the maturation of mitochondrial 4Fe-4S proteins functioning late in the iron-sulfur cluster assembly pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf69
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IBA57
Gene Accession No. Q5T440, Q8CAK1
Gene Alias 4930543L23Rik; A230051G13Rik; C1orf69; IBA57; IBA57 homolog, iron-sulfur cluster assembly; IBA57, iron-sulfur cluster assembly homolog; IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae); iron-sulfur cluster assembly factor for biotin synthase- and aconitase-like mitochondrial proteins, with a mass of 57kDa; iron-sulfur cluster assembly factor homolog; MMDS3; putative transferase C1orf69 homolog, mitochondrial; putative transferase C1orf69, mitochondrial; putative transferase CAF17 homolog, mitochondrial; Putative transferase CAF17, mitochondrial; RGD1306000; SPG74
Gene Symbols IBA57
Host Species Rabbit
Immunogen Recombinant Human Putative transferase CAF17, mitochondrial protein (286-356AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200205, 216792
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.