Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531680 Supplier Invitrogen™ Supplier No. PA531680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

C1orf74, also known as URCL4, is located on chromosome 1q32.2 and encodes a previously uncharacterized protein. Recent studies have revealed that C1orf74 is significantly up-regulated in various cancers, including cervical cancer and lung adenocarcinoma. The overexpression of C1orf74 is associated with poor prognosis, indicating its potential role in tumor progression and proliferation. Mechanistically, C1orf74 has been shown to positively regulate the EGFR/AKT/mTORC1 signaling pathway, which is crucial for cell growth and survival. Knockdown studies have demonstrated that reducing C1orf74 expression inhibits cancer cell growth, highlighting it as a potential therapeutic target. Additionally, C1orf74's role in immune cell infiltration suggests it may influence the tumor immune microenvironment, further implicating its importance in cancer biology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf74
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene C1orf74
Gene Accession No. Q96LT6
Gene Alias C1orf74; chromosome 1 open reading frame 74; UPF0739 protein C1orf74; up-regulated in lung cancer 4; URLC4
Gene Symbols C1orf74
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C1orf74. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148304
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.