Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf77 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544307 Supplier Invitrogen™ Supplier No. PA544307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LKQSLKQRLG KSNIQARLGR PIGALARGAI GGRGLPIIQR GLPRGGLRGG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100% PA5-44307 detects C1orf77 in IF application with mouse samples.

CHTOP is a small nuclear protein that is characterized by an arginine and glycine rich region. This protein may have an important role in the regulation of fetal globin gene expression and in the activation of estrogen-responsive genes. A recent study reported that this protein binds 5-hydroxymethylcytosine (5hmC) and associates with an arginine methyltransferase complex (methylosome), which promotes methylation of arginine 3 of histone H4 (H4R3) and activation of genes involved in glioblastomagenesis. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf77
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHTOP
Gene Accession No. Q9CY57, Q9Y3Y2
Gene Alias 2500003M10Rik; C1orf77; C3H1orf77; chromatin target of PRMT1; chromatin target of PRMT1 protein; Chtop; DKFZP547E1010; FL-SRAG; FOP; Friend of Prmt1; Friend of PRMT1 protein; HT031; hypothetical protein LOC616638; MNCb-1706; PP7704; RP1-178F15.2; similar to DKFZP547E1010 protein; small arginine- and glycine-rich protein; small protein rich in arginine and glycine; Srag; SRAG-3; SRAG-5
Gene Symbols CHTOP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C1orf77.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26097, 66511
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.