Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144258
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144258 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144258 Supplier Invitrogen™ Supplier No. PA5144258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf84 Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf84
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SZT2
Gene Accession No. Q5T011
Gene Alias 6430407N22; C1orf84; EIEE18; ENSMUSG00000058432; KIAA0467; KICSTOR complex protein SZT2; protein SZT2; RGD1308616; RP11-506B15.1; RP11-506B15.3; seizure threshold 2; seizure threshold 2 homolog (mouse); Seizure threshold 2 protein; Seizure threshold 2 protein homolog; Szt2; SZT2A; SZT2B; TIGR; Transcript increased in glutamate resistance
Gene Symbols SZT2
Host Species Rabbit
Immunogen Recombinant Human Protein SZT2 protein (2396-2517AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23334
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.