Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf84 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983429
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3429 Supplier Bioss Supplier No. BS15077R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C1orf84
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SZT2
Gene Accession No. Q5T011
Gene Alias C1orf84; EIEE18; KIAA0467; KICSTOR complex protein SZT2; protein SZT2; RP11-506B15.1; RP11-506B15.3; seizure threshold 2 homolog (mouse); Seizure threshold 2 protein homolog; SZT2; SZT2A; SZT2B
Gene Symbols SZT2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf84.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23334
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.