Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf85 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142985
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142985 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142985 Supplier Invitrogen™ Supplier No. PA5142985
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: The immunizing peptide represents the luminal part of the protein. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf85
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GLMP
Gene Accession No. Q68FV6, Q9JHJ3
Gene Alias 0610031j06rik; AB027141; C1orf85; GLMP; Glycosylated lysosomal membrane protein; kidney lysosomal membrane protein; kidney predominant protein NCU-G1; lysosomal protein NCU-G1; NCU-G1; PSEC0030; RGD1303130; UNQ2553/PRO6182
Gene Symbols GLMP
Host Species Goat
Immunogen Peptide with sequence PDCPSVNERNSIDD (aa 243-256).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295231, 56700
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.