Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf86 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546347 Supplier Invitrogen™ Supplier No. PA546347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ARSLPQRPAP DPCRAPRVEQ QPSVEGAAAL RSCPMCQKEF APRLTQLDVD Sequence homology: Human: 100%.

C1orf86 is a component of the Fanconi anemia (FA) complex required to recruit the FA complex to DNA interstrand cross-links (ICLs) and promote ICLs repair. Following DNA damage recognizes and binds 'Lys-63'-linked ubiquitin generated by RNF8 at ICLs and recruits other components of the FA complex. It promotes translesion synthesis via interaction with REV1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf86
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAAP20
Gene Accession No. Q6NZ36
Gene Alias 2610002J02Rik; C1orf86; Faap20; FANCA-associated protein of 20 kDa; Fanconi anemia core complex associated protein 20; Fanconi anemia core complex-associated protein 20; Fanconi anemia-associated protein of 20 kDa; FP7162; likely orthologue of H. sapiens chromosome 1 open reading frame 86 (C1orf86); LOC362678; RGD1308923; RIKEN cDNA 2610002J02 gene
Gene Symbols FAAP20
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C1orf86 (aa 115-164).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199990
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.