Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIFO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571443
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571443 Supplier Invitrogen™ Supplier No. PA571443
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIFO Polyclonal Antibody for Western Blot

During primary cilia disassembly, involved in cilia disassembly. Required specifically to control cilia retraction as well as the liberation and duplication of the basal body/centrosome. May act by stimulating AURKA activity at the basal body in a cell cycle-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIFO
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PIFO
Gene Accession No. Q8TCI5
Gene Alias C1orf88; PIFO; pitchfork; primary cilia formation; protein pitchfork
Gene Symbols PIFO
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C1ORF88.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128344
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.