Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf96 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560366 Supplier Invitrogen™ Supplier No. PA560366
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEQDAEAGDA EAEDAEDAAL PALPVKDVED KPEQQTRTRE TDKSPTSTEP RQQPSALFAR GNRKAVKSPQ RSSSKIKENK HPFALYGW Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.

Plays a role in microtubule (MT) stabilization and this stabilization involves the maintenance of NUMA1 at the spindle poles. Colocalizes with polyglutamylated MTs to promote MT stabilization and regulate bipolar spindle formation in mitosis. Binding of CCSAP to centrosomes and the spindle around centrosomes during mitosis inhibits MT depolymerization, thereby stabilizing the mitotic spindle (PubMed:26562023). May play a role in embryonic development. May be required for proper cilia beating. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf96
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCSAP
Gene Accession No. Q6IQ19
Gene Alias 1700054N08Rik; AI853424; C1orf96; C4H1orf96; Ccsap; centriole and spindle-associated protein; centriole, cilia and spindle associated protein; centriole, cilia and spindle-associated protein; CSAP; RGD1563235
Gene Symbols CCSAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf96. Recombinant protein control fragment (Product #RP-94329).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.