Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf96 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543904 Supplier Invitrogen™ Supplier No. PA543904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LGRRLLEQAH APWLWDDWGP AGSSEDSASS ESSGAGGPAP RCAPPSPPPP Sequence homology: Dog: 79%; Human: 100%.

Plays a role in microtubule (MT) stabilization and this stabilization involves the maintenance of NUMA1 at the spindle poles. Colocalizes with polyglutamylated MTs to promote MT stabilization and regulate bipolar spindle formation in mitosis. Binding of CCSAP to centrosomes and the spindle around centrosomes during mitosis inhibits MT depolymerization, thereby stabilizing the mitotic spindle (PubMed:26562023). May play a role in embryonic development. May be required for proper cilia beating. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf96
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCSAP
Gene Accession No. Q6IQ19
Gene Alias 1700054N08Rik; AI853424; C1orf96; C4H1orf96; Ccsap; centriole and spindle-associated protein; centriole, cilia and spindle associated protein; centriole, cilia and spindle-associated protein; CSAP; RGD1563235
Gene Symbols CCSAP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C1orf96 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126731
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.