Learn More
Invitrogen™ C1QA Polyclonal Antibody

Description
Immunogen sequence: EDLCRAPDGK KGEAGRPGRR GRPGLKGEQG EPGAPGIRTG IQGLKGDQGE PGPSGNPGKV GYPGPSGPLG ARGIPGIKGT KGSPGNIKDQ PRPAFSAIRR NPPMGGNVVI FDTVITNQEE PYQNHSGRFV CTVPGYYYFT FQVLSQWEIC LSIVSSSRGQ VRRSLGFCDT TNKGLFQVVS GGMVLQLQQG DQVWVEKDPK KGHIYQGSEA DSVFSGFLIF PSA; Positive Samples: Mouse lung, Mouse liver.
Specifications
Specifications
| Antigen | C1QA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | C1qa |
| Gene Accession No. | P02745, P31720, P98086 |
| Gene Alias | AI255395; C1q; C1QA; complement C1q A chain; complement C1q chain A; complement C1q subcomponent subunit A; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component C1q, A chain |
| Gene Symbols | C1qa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.