Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1q Polyclonal Antibody, FITC

Catalog No. PIPA184593
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA184593 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA184593 Supplier Invitrogen™ Supplier No. PA184593

Sheep Polyclonal Antibody

Sheep anti Human C1q antibody recognizes human C1q, a sub-component of the C1 complement component.

C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1q
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 10 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene C1qc
Gene Accession No. P02745, P02746, P02747
Gene Alias AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B
Gene Symbols C1QA, C1QB, C1qc
Host Species Sheep
Immunogen Human C1q, purified from plasma.
Purification Method Ion-exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 712, 713, 714
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.