Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1q Antibody, Novus Biologicals™
SDP

Catalog No. NB119898 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119898 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119898 Supplier Novus Biologicals Supplier No. NBP311273
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1q Polyclonal specifically detects C1q in Rat samples. It is validated for Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen C1q
Applications Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias A chain, complement component 1, q subcomponent, A chain, complement component 1, q subcomponent, alpha polypeptide
Host Species Rabbit
Immunogen The polyclonal antibody recognizes rat C1q.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 712
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.