Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QB Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299263AF488
Description
C1QB Polyclonal specifically detects C1QB in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C1QB | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse C1QB. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
713 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction