Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    C1QB Antibody (C1QB/2966), DyLight 594, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279922DL594
Description
C1QB Monoclonal specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| C1QB | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C1QB | |
| Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) (Uniprot: P02746) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | 
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| C1QB/2966 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| IgG1 κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction