Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QB Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299263MFV450
Description
C1QB Polyclonal antibody specifically detects C1QB in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
C1QB | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C1QB | |
0.1 mL | |
Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration, Neuroscience | |
713 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction