Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QB Antibody (SR1418), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32186850UL
Description
C1QB Monoclonal antibody specifically detects C1QB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, SerumSpecifications
C1QB | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1418 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Serum 1:50-1:200 | |
B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
A synthesized peptide derived from human C1QB (Uniprot #: P02746) | |
50 μg | |
Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration, Neuroscience | |
713 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction