Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QB Antibody (C1QB/2966), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279922AF700
Description
C1QB Monoclonal specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C1QB | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C1QB | |
Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) (Uniprot: P02746) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
C1QB/2966 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction