Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109461 Supplier Invitrogen™ Supplier No. PA5109461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGAPGPKGES GDYKATQKIA FSATRTINVP LRRDQTIRFD HVITNMNNNY EPRSGKFTCK VPGLYYFTYH ASSRGNLCVN LMRGRERAQK VVTFCDYAYN TFQVTTGGMV LKLEQGENVF LQATDKNSLL GMEGANSIFS GFLLFPDMEA.

C1QB is a major constituent of the human complement subcomponent C1q. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene C1QB
Gene Accession No. P14106
Gene Alias C1QB; C1QTNF5; complement C1q B chain; complement C1q chain B; complement C1q subcomponent subunit B; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component C1q, B chain; complement subcomponent C1q chain B; CTRP5; DKFZP586B0621; LORD
Gene Symbols C1QB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 104-253 of human C1QB (NP_000482.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12260
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.