Learn More
Invitrogen™ C1QBP Monoclonal Antibody (60.11), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548988
Description
The monoclonal antibody 60.11 is directed against epitopes situated within the NH2-terminal stretch of gC1q-R corresponding to residues 76-93. Clone 60.11 recognizes the putative C1q binding site and reacts with the mature form, but has poor or no reactivity with the truncated form, lacking residues 74-95.
Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.
Specifications
C1QBP | |
Monoclonal | |
100 μg/mL | |
PBS with 1% BSA and 0.02% sodium azide | |
Q07021 | |
C1QBP | |
Recombinant GC1q-R corresponding to mature GC1q-R (amino acids 74-282). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Inhibition Assays, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
60.11 | |
FITC | |
C1QBP | |
AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein | |
Mouse | |
Protein G | |
RUO | |
708 | |
4°C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.