Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1QBP Monoclonal Antibody (74.5.2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548989
Description
The monoclonal antibody 74.5.2 is directed against epitopes in the XC15 peptide that contains a binding site for high-molecular-weight kininogen and Factor XII. Clone 74.5.2 recognizes both the mature (74-282) and truncated form, lacking residues 74-95.
Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.
Specifications
C1QBP | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and no preservative | |
Q07021 | |
C1QBP | |
Recombinant gC1q-R corresponding to mature GC1q-R (amino acids 74-282). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Inhibition Assays, Immunoprecipitation, Western Blot, Immunocytochemistry | |
74.5.2 | |
Unconjugated | |
C1QBP | |
AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein | |
Mouse | |
Protein G | |
RUO | |
708 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction