Learn More
Invitrogen™ C1QBP Recombinant Rabbit Monoclonal Antibody (6N7U3)

Description
Immunogen sequence: KTLPKMSGGW ELELNGTEAK LVRKVAGEKI TVTFNINNSI PPTFDGEEEP SQGQKVEEQE PELTSTPNFV VEVIKNDDGK KALVLDCHYP EDEVGQEDEA E.
Specifications
Specifications
| Antigen | C1QBP |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 6N7U3 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | C1QBP |
| Gene Accession No. | O35658, O35796, Q07021 |
| Gene Alias | AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.