Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595975 Supplier Invitrogen™ Supplier No. PA595975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHTDGDKAFV DFLSDEIKEE RKIQKHKTLP KMSGGWELEL NGTEAKLVRK VAGEKITVTF NINNSIPPTF DGEEEPSQGQ KVEEQEPELT STPNFVVEVI KNDDGKKALV LDCHYPEDEV GQEDEAESDI FSIREVSFQS TGESEWKDTN YTLNTDSLDW ALYDHLMDFL ADRGVDNTFA DELVELSTAL EHQEYITFLE DLKSFVKSQ; Positive Samples: CEM, SW620, HeLa, Jurkat, Mouse liver, Mouse kidney, Mouse heart, Mouse intestine, Rat brain; Cellular Location: Cell membrane, Cytoplasm, Extracellular side, Mitochondrion matrix, Nucleus, Peripheral membrane protein, nucleolus.

Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QBP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C1QBP
Gene Accession No. O35658, O35796, Q07021
Gene Alias AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein
Gene Symbols C1QBP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 74-282 of human GC1q R/GC1q R/C1QBP (NP_0012031).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12261, 29681, 708
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.