Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1QC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551583
Description
Immunogen sequence: GIPGEPGEEG RYKQKFQSVF TVTRQTHQPP APNSLIRFNA VLTNPQGDYD TSTGKFTCKV PGLYYFVYHA SHTANLCVLL YRSGVKVVTF CGHTSKTNQV NSGGVLLRLQ VGEEVWLAVN DYYDMVGIQG SD Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.
This gene encodes a major constituent of the human complement subcomponent C1q. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. A deficiency in C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N-terminus, and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1. This gene encodes the C-chain polypeptide of human complement subcomponent C1q. Alternatively spliced transcript variants that encode the same protein have been found for this gene.
Specifications
C1QC | |
Polyclonal | |
Unconjugated | |
C1QC | |
AI385742; C1qc; C1Q-C; C1qg; Ciqc; complement C1q C chain; complement C1q subcomponent subunit C; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
714 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P02747 | |
C1QC | |
Recombinant protein corresponding to Human C1QC. Recombinant protein control fragment (Product #RP-95389). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction