Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1ql1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569825 Supplier Invitrogen™ Supplier No. PA569825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

C1QL1 gene ontology annotations related to this gene include maintenance of synapse structure; motor learning; neuron remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1ql1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C1QL1
Gene Accession No. O88992
Gene Alias C1q and tumor necrosis factor-related protein 14; C1q related factor; C1q/TNF-related protein 14; C1ql1; C1q-related factor; C1QRF; C1QTNF14; complement C1q like 1; Complement component 1 Q subcomponent-like 1; complement component 1, q subcomponent-like 1; CRF; CTRP14; gliacolin; LOW QUALITY PROTEIN: C1q-related factor
Gene Symbols C1QL1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse C1ql1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23829
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.