Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101536 Supplier Invitrogen™ Supplier No. PA5101536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C1QL2.

May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C1QL2
Gene Accession No. Q7Z5L3, Q8CFR0
Gene Alias BC040774; C1q and tumor necrosis factor related protein 10; C1q and tumor necrosis factor-related protein 10; C1q/TNF-related protein 10; C1q-domain containing protein; C1QL2; C1qtnf10; complement C1q like 2; complement C1q-like protein 2; complement component 1, q subcomponent-like 2; Ctrp10
Gene Symbols C1QL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human C1QL2(Accession Q7Z5L3), corresponding to amino acid residues A231-G281.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165257, 226359
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.