Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570866 Supplier Invitrogen™ Supplier No. PA570866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1QL3 Polyclonal Antibody for Western Blot

May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses. Plays a role in glucose homeostasis. Via AMPK signaling pathway, stimulates glucose uptake in adipocytes, myotubes and hepatocytes and enhances insulin-stimulated glucose uptake. In a hepatoma cell line, reduces the expression of gluconeogenic enzymes G6PC and PCK1 and hence decreases de novo glucose production. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C1QL3
Gene Accession No. Q5VWW1
Gene Alias 1110065A22Rik; AI661623; C1q and tumor necrosis factor-related protein 13; C1q/TNF-related protein 13; C1ql; C1ql3; C1q-like 3; C1qtnf13; complement C1q like 3; complement C1q-like protein 3; complement component 1, q subcomponent-like 3; CTRP13; Gliacolin; K100
Gene Symbols C1QL3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C1QL3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389941
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.