Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103759 Supplier Invitrogen™ Supplier No. PA5103759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C1QL3.

May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses. Plays a role in glucose homeostasis. Via AMPK signaling pathway, stimulates glucose uptake in adipocytes, myotubes and hepatocytes and enhances insulin-stimulated glucose uptake. In a hepatoma cell line, reduces the expression of gluconeogenic enzymes G6PC and PCK1 and hence decreases de novo glucose production. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C1QL3
Gene Accession No. Q5VWW1, Q9ESN4
Gene Alias 1110065A22Rik; AI661623; C1q and tumor necrosis factor-related protein 13; C1q/TNF-related protein 13; C1ql; C1ql3; C1q-like 3; C1qtnf13; complement C1q like 3; complement C1q-like protein 3; complement component 1, q subcomponent-like 3; CTRP13; Gliacolin; K100
Gene Symbols C1QL3
Host Species Rabbit
Immunogen A synthesized peptide derived from human C1QL3(Accession Q5VWW1), corresponding to amino acid residues L110-T160.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227580, 389941
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.