Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1qR1/CD93 Antibody (223437) [FITC], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1696F
Description
C1qR1/CD93 Monoclonal antibody specifically detects C1qR1/CD93 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
C1qR1/CD93 | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
223437 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
Mouse myeloma cell line NS0-derived recombinant mouse C1q R1/CD93, Ala23-Asn572, Accession # O89103 | |
0.1 mL | |
B Cell Development and Differentiation Markers | |
22918 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction