Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1qR1/CD93 Antibody (273107), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB23791J
Description
C1qR1/CD93 Monoclonal antibody specifically detects C1qR1/CD93 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
C1qR1/CD93 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
273107 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
Mouse myeloma cell line NS0-derived recombinant human C1q R1/CD93, Ala24-Lys580, Accession # Q9NPY3 | |
0.1 mL | |
B Cell Development and Differentiation Markers | |
22918 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction