Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QTNF10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989352
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9352 Supplier Bioss Supplier No. BS9793R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses.

Specifications

Antigen C1QTNF10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C1QL2
Gene Accession No. Q7Z5L3
Gene Alias BC040774; C1q and tumor necrosis factor related protein 10; C1q and tumor necrosis factor-related protein 10; C1q/TNF-related protein 10; C1q-domain containing protein; C1ql2; C1qtnf10; complement C1q like 2; Complement C1q-like protein 2; complement component 1, q subcomponent-like 2; Ctrp10
Gene Symbols C1QL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1QL2/C1QTNF10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165257
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.