Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1qTNF5/CTRP5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176631SS
Description
C1qTNF5/CTRP5 Polyclonal specifically detects C1qTNF5/CTRP5 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C1qTNF5/CTRP5 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3 | |
Rabbit | |
Affinity Purified | |
Vision | |
114902 | |
Human, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
AAQ88749 | |
C1QTNF5 | |
Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human CTRP5. The immunogen is located within amino acids 30 - 80 of CTRP5. Amino Acid Squence: LPGREDGREGREGAPG | |
0.025 mg | |
Primary | |
These proteins are often highly modified post-translationally and migrate in SDS-PAGE at positions other than their predicted size. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction