Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QTNF5, Rabbit anti-Human, Clone: A14-A, Abnova™

Catalog No. 89998739 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-739 Supplier Abnova Corporation Supplier No. MAB15899
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen C1QTNF5
Applications ELISA, Western Blot
Classification Monoclonal
Clone A14-A
Conjugate Unconjugated
Dilution ELISA (1:50000-1:100000) Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide).
Gene C1q and tumor necrosis factor related protein 5
Gene Alias CTRP5/DKFZp586B0621/LORD
Gene Symbols C1QTNF5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 109-121 at N-terminus of human C1QTNF5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114902
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.