Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QTNF6, Mouse anti-Human, Clone: 256-E, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16078
Specifications
C1QTNF6 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4. | |
CTRP6/ZACRP6 | |
Mouse | |
100 μg | |
Primary | |
Recognizes the N-terminal half of human C1QTNF6 full-length protein. | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
256-E | |
ELISA (1:200-500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:200-500) The optimal working dilution should be determined by the end user. | |
C1q and tumor necrosis factor related protein 6 | |
C1QTNF6 | |
Recombinant protein corresponding to globular domain-less of human C1QTNF6. | |
RUO | |
114904 | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction