Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QTNF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570867 Supplier Thermo Scientific Supplier No. PA570867
Add to Cart
Add to Cart

C1QTNF8 Polyclonal Antibody

C1QTNF8 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QTNF8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C1QTNF8
Gene Accession No. P60827
Gene Alias C1q and TNF related 8; C1q and tumor necrosis factor related protein 8; C1q/TNF-related protein 8; C1QTNF8; Complement C1q tumor necrosis factor-related protein 8; CTRP8; UNQ5829; UNQ5829/PRO19648
Gene Symbols C1QTNF8
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human C1QTNF8.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390664
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.