Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QTNF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103757 Supplier Invitrogen™ Supplier No. PA5103757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C1QTNF8.

C1QTNF8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QTNF8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C1QTNF8
Gene Accession No. P60827
Gene Alias C1q and TNF related 8; C1q and tumor necrosis factor related protein 8; C1q/TNF-related protein 8; C1QTNF8; Complement C1q tumor necrosis factor-related protein 8; CTRP8; UNQ5829; UNQ5829/PRO19648
Gene Symbols C1QTNF8
Host Species Rabbit
Immunogen A synthesized peptide derived from human C1QTNF8(Accession P60827), corresponding to amino acid residues R199-A249.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390664
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.