Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QTNF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121392 Supplier Invitrogen™ Supplier No. PA5121392
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

Probable adipokine. Activates AMPK, AKT, and p44/42 MAPK signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QTNF9
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene C1QTNF9
Gene Accession No. P0C862
Gene Alias 9130217G22Rik; AQL1; C1q and tumor necrosis factor related protein 9; C1QTNF9; C1QTNF9A; Ciqtnf9; Complement C1q and tumor necrosis factor-related protein 9; complement C1q and tumor necrosis factor-related protein 9A; complement C1q tumor necrosis factor-related protein 9; complement C1q tumor necrosis factor-related protein 9A; CTRP9; RGD1560876; UNQ6503/PRO21380
Gene Symbols C1QTNF9
Host Species Rabbit
Immunogen Recombinant Human Complement C1q and tumor necrosis factor-related protein 9A protein (58-72aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338872
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.