Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1QTNF9 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15085R

Specifications
C1QTNF9 | |
Polyclonal | |
Unconjugated | |
C1QTNF9 | |
AQL1; C1q and tumor necrosis factor related protein 9; C1QTNF9; C1QTNF9A; Complement C1q and tumor necrosis factor-related protein 9; Complement C1q and tumor necrosis factor-related protein 9A; complement C1q tumor necrosis factor-related protein 9; complement C1q tumor necrosis factor-related protein 9A; CTRP9; UNQ6503/PRO21380 | |
Rabbit | |
Protein A | |
RUO | |
338872 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P0C862 | |
C1QTNF9 | |
KLH conjugated synthetic peptide derived from human C1QTNF9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction