Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1QTNF9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103758 Supplier Invitrogen™ Supplier No. PA5103758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C1QTNF9B.

Probable adipokine. Activates AMPK, AKT, and p44/42 MAPK signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QTNF9B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C1QTNF9B
Gene Accession No. B2RNN3
Gene Alias C1q and tumor necrosis factor related protein 9B; C1q/TNF-related protein 9B; C1QTNF9B; collagen triple helix repeat-containing; Complement C1q and tumor necrosis factor-related protein 9B; Complement C1q and tumor necrosis factor-related protein 9-like; CTRP9B
Gene Symbols C1QTNF9B
Host Species Rabbit
Immunogen A synthesized peptide derived from human C1QTNF9B(Accession B2RNN3), corresponding to amino acid residues Y253-L303.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387911
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.