Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1r Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576140
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
C1 component of human complement is a complex of three glycoproteins: C1q, C1r and C1s. C1r has a molecular weight of 83,000 and is present in plasma in an average concentration of 50 μg/mL, either as a complex with C1q and C1s or in the absence of C1q as a tetramer with C1s. The subcomponents of the C1 complex are responsible for the initiation of the classical pathway of complement activation.
Specifications
C1r | |
Polyclonal | |
Unconjugated | |
C1R | |
AI132558; C1r; C1ra; complement C1r; complement C1r subcomponent; Complement C1r subcomponent heavy chain; Complement C1r subcomponent light chain; Complement C1r-A subcomponent; Complement C1r-A subcomponent heavy chain; Complement C1r-A subcomponent light chain; Complement component 1 subcomponent r; Complement component 1 subcomponent r-A; complement component 1, r subcomponent; complement component 1, r subcomponent A; complement component 1, r-A subcomponent; mC1rA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
312705, 50909, 715 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
P00736, Q8CG16 | |
C1R, C1ra | |
Recombinant full length Human C1r (LC). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction