Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1r Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109463 Supplier Invitrogen™ Supplier No. PA5109463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGQKAKMGNF PWQVFTNIHG RGGGALLGDR WILTAAHTLY PKEHEAQSNA SLDVFLGHTN VEELMKLGNH PIRRVSVHPD YRQDESYNFE GDIALLELEN SVTLGPNLLP ICLPDNDTFY DLGLMGYVSG FGVMEEKIAH DLRFVRLPVA NPQACENWLR GKNRMDVFSQ NMFCAGHPSL KQDACQGDSG GVFAVRDPNT DRWVATGIVS WGIGCSRGYG FYTKVLNYVD WIKKEMEEED.

C1 component of human complement is a complex of three glycoproteins: C1q, C1r and C1s. C1r has a molecular weight of 83,000 and is present in plasma in an average concentration of 50 μg/mL, either as a complex with C1q and C1s or in the absence of C1q as a tetramer with C1s. The subcomponents of the C1 complex are responsible for the initiation of the classical pathway of complement activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1r
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1.90 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene C1R
Gene Accession No. P00736
Gene Alias AI132558; C1r; C1ra; complement C1r; complement C1r subcomponent; Complement C1r subcomponent heavy chain; Complement C1r subcomponent light chain; Complement C1r-A subcomponent; Complement C1r-A subcomponent heavy chain; Complement C1r-A subcomponent light chain; Complement component 1 subcomponent r; Complement component 1 subcomponent r-A; complement component 1, r subcomponent; complement component 1, r subcomponent A; complement component 1, r-A subcomponent; mC1rA
Gene Symbols C1R
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 466-705 of human C1R (NP_001724.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 715
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.