Learn More
Invitrogen™ C1rLP Monoclonal Antibody (393608)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) C1r, rhC1s, rhHaptoglobin, or rhMASP3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | C1rLP |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 393608 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | C1RL |
| Gene Accession No. | Q9NZP8 |
| Gene Alias | C1RL; C1RL1; C1r-like protein; C1r-like serine protease analog protein; C1RLP; C1r-LP; CLSPA; complement C1r subcomponent like; complement C1r subcomponent-like protein; complement C1r-like proteinase; complement component 1, r subcomponent like protein; complement component 1, r subcomponent-like |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.