Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1rLP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582430 Supplier Invitrogen™ Supplier No. PA582430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVLPVCLPDN ETLYRSGLLG YVSGFGMEMG WLTTELKYSR LPVAPREACN AWLQKRQRPE VFSDNMFCVG DETQ Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 84%.

Complement component 1, r subcomponent-like protein (C1rLP) is part of the complement system, primarily involved in innate immunity. Located on human chromosome 12p13.31, C1rLP is closely related to C1r, a serine protease component of the C1 complex in the classical pathway of complement activation. C1rLP shares structural similarities with C1r, featuring serine protease domains that are crucial for the proteolytic processing required to activate the complement cascade. The precise functional role of C1rLP remains under investigation, but it is hypothesized to act as a modulator of complement activation, potentially influencing immune cell behavior during inflammation or infection. The potential existence of several isoforms of C1rLP suggests diverse regulatory functions, which could impact processes like opsonization, pathogen clearance, and inflammation. Current research explores its relevance in immune response regulation, with implications for understanding autoimmune and inflammatory diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1rLP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1RL
Gene Accession No. Q9NZP8
Gene Alias C1RL; C1RL1; C1r-like protein; C1r-like serine protease analog protein; C1RLP; C1r-LP; CLSPA; complement C1r subcomponent like; complement C1r subcomponent-like protein; complement C1r-like proteinase; complement component 1, r subcomponent like protein; complement component 1, r subcomponent-like
Gene Symbols C1RL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1RL. Recombinant protein control fragment (Product #RP-89272).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.